missing translation for 'onlineSavingsMsg'
Learn More

Serdemetan, MedChemExpress

Product Code. 16464928
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16464928 10 mg 10mg
16455706 100 mg 100mg
16431177 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16464928 Supplier Medchem Express Supplier No. HY12025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Serdemetan(JNJ-26854165) acts as a HDM2 ubiquitin ligase antagonist and also induces early apoptosis in p53 wild-type cells, inhibits cellular proliferation followed by delayed apoptosis in the absence of functional p53.

Serdemetan(JNJ-26854165) acts as a HDM2 ubiquitin ligase antagonist and also induces early apoptosis in p53 wild-type cells, inhibits cellular proliferation followed by delayed apoptosis in the absence of functional p53.

Specifications

Chemical Name or Material Serdemetan
CAS 881202-45-5
Color Brown
Purity Grade Notes Research
Molecular Formula C21H20N4
Quantity 10 mg
Synonym JNJ-26854165
Solubility Information DMSO : 50 mg/mL (152.25 mM; Need ultrasonic)
SMILES C1(NCCC2=CNC3=C2C=CC=C3)=CC=C(C=C1)NC4=CC=NC=C4
Molecular Weight (g/mol) 328.41
Formula Weight 328.41
Percent Purity 99.23%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.