Learn More
Applied Biosystems™ SeqStudio™ Sequence Analysis Software
Product Code. 15863540 Shop All Applied Biosystems Products
Change view
Click to view available options
For Use With (Equipment):
SeqStudio™ Genetic Analyzer
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) unitSize
15863540 SeqStudio™ Genetic Analyzer Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15863540 Supplier Applied Biosystems™ Supplier No. 4443764
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
Sequence Analysis Software for SeqStudio™
Sequence Analysis Software for SeqStudio™
Order Info
Each
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage 2 to 8°C
For Use With (Equipment) SeqStudio™ Genetic Analyzer
Product Type Sequence Analysis Software