Learn More
Fisherbrand™ Septum for Magnetic Universal Precision Thread Cap
Product Code. 11805000 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Color:
Gray, Red
Unit Size:
Pack of 1000
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
11805000 Gray, Red Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11805000
Estimated Shipment: 14-12-2026
Add to basket
Add to basket
Most septa are laminated on one side with PTFE for high chemical resistance and an inert barrier between sample and carrier material.
  • 17mm Septa
TRUSTED_SUSTAINABILITY
Specifications
Description Septa
Color Gray, Red
Diameter (Metric) Outer 17 mm
Material Butyl, PTFE
Quantity 1000
Thickness (Metric) 1.6 mm
For Use With (Equipment) Magnetic Universal Precision Thread Cap
Product Type Septa