Learn More
Fisherbrand™ Septum for 24-400 Screw Thread Cap
Product Code. 11835070 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Color:
Blue, White
Gray, Red
Red, White
Red-Orange, Transparent
Silver, White
Material:
Aluminum, Silicone
Butyl, PTFE
Natural Rubber, TEF
PTFE, Silicone
Thickness (Metric):
1.3 mm
1.5 mm
1.6 mm
2.5 mm
3 mm
Unit Size:
Pack of 1000
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Material Thickness (Metric) Unit Size
11835070 Gray, Red Butyl, PTFE 1.6 mm Pack of 1000
11845070 Gray, Red Butyl, PTFE 2.5 mm Pack of 1000
11815070 Blue, White PTFE, Silicone 1.5 mm Pack of 1000
11825070 Red-Orange, Transparent Natural Rubber, TEF 1.3 mm Pack of 1000
16328387 Silver, White Aluminum, Silicone 3 mm Pack of 1000
12635887 Red, White PTFE, Silicone 1.3 mm Pack of 1000
15397827 Blue, White PTFE, Silicone 1.5 mm Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Product Code. 11835070
Save Up to 60% on Contract Testing Essentials
Use promo code 28025
View all promotions
Estimated Shipment: 02-10-2026
Add to basket
Add to basket
Most septa are laminated on one side with PTFE for high chemical resistance and an inert barrier between sample and carrier material.
  • 22mm Septa
TRUSTED_SUSTAINABILITY
Specifications
Description Septa
Color Gray, Red
Diameter (Metric) Outer 22 mm
Material Butyl, PTFE
Quantity 1000
Thickness (Metric) 1.6 mm