Learn More
Fisherbrand™ Septum for 13-425 screw cap
Product Code. 11894950 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Color:
Blue Transparent, White
Blue, White
Cream, Red
Gray, Red
Red, White
Red-Orange, Transparent
White
Material:
Butyl, PTFE
Natural Rubber, TEF
PTFE Virginal
PTFE, Silicone
Thickness (Metric):
0.25 mm
1 mm
1.3 mm
1.5 mm
Unit Size:
Pack of 1000
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Material Thickness (Metric) Unit Size
11894950 Blue Transparent, White PTFE, Silicone 1.3 mm Pack of 1000
11834950 Red-Orange, Transparent Natural Rubber, TEF 1 mm Pack of 1000
11757597 Gray, Red Butyl, PTFE 1.3 mm Pack of 1000
11814950 Red-Orange, Transparent Natural Rubber, TEF 1.3 mm Pack of 1000
11824950 Blue, White PTFE, Silicone 1.5 mm Pack of 1000
11894940 Cream, Red PTFE, Silicone 1.5 mm Pack of 1000
11804950 White PTFE Virginal 0.25 mm Pack of 1000
11844950 Red, White PTFE, Silicone 1 mm Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Product Code. 11894950
Big Savings for Better Separation
Use promo code 27992
View all promotions
Estimated Shipment: 10-12-2026
Add to basket
Add to basket
Most septa are laminated on one side with PTFE for high chemical resistance and an inert barrier between sample and carrier material.
  • 12mm Septa
TRUSTED_SUSTAINABILITY
Specifications
Description Septa
Color Blue Transparent, White
Diameter (Metric) Outer 12 mm
Material PTFE, Silicone
Quantity 1000
Thickness (Metric) 1.3 mm