Learn More
Mettler Toledo™ Sensor Holder, EasyPlace, Triple
Shop All Mettler Toledo Products
Click to view available options
For Use With (Equipment):
EasyPlace
Unit Size:
Each
Description
- Three-in-One
- For multi-channel measurements, external references or temperature sensors - position multiple sensor with one EasyPlace
Specifications
Specifications
| Type | Sensor Holder, EasyPlace Triple |
| For Use With (Equipment) | EasyPlace |