Learn More
Thermo Scientific™ Sensititre™ EU Surveillance Enterococcus EUVENC AST Plate
Descripción
Comply with EU Surveillance System program for Enterococcus species with this convenient plate.
Perform accurate AST with Enterococcus species-specific formulary as a part of EU Surveillance program with Thermo Scientific™ Sensititre™ EU Surveillance Enterococcus EUVENC AST Plate.
Comply with EU Surveillance System program for Enterococcus species with Sensititre EU Surveillance Enterococcus EUVENC AST Plate
- Extended shelf life, room temperature storage eliminates inventory control concerns
- Individual packaging allows a laboratory to test one plate at a time with no waste
- Inclusive on-scale QC ranges provide immediate quality assurance of testing methodology
- Automated or manual read options are available for ultimate flexibility
Antimicrobial Resistance (AMR) is a significant concern and of increasing importance. The inappropriate use of antimicrobial agents in humans and animals is leading this growing and dangerous issue, emerging as a public health threat. This is why the European Parliament, the Council and its Agencies: European Centre for Disease Prevention and Control (ECDC), the European Food Safety Authority (EFSA) and the European Medicines Agency (EMA), have identified the need to establish a common strategy in the fight against AMR.
The Sensititre System utilizes true minimum inhibitory concentration (MIC) results, which are essential to identifying antimicrobial resistance, and provide greater sensitivity for better resistance tracking.
With a wide range of Sensititre plates specifically designed for surveillance programs, the Sensititre System is a leader in antimicrobial susceptibility testing for surveillance.
Not all products are available for sale in all territories. Please inquire.
VersaTREK™ and Sensititre™ products are now part of the Thermo Scientific brand
Especificaciones
Especificaciones
| Description | Sensititre EU Surveillance Enterococcus EUVENC AST Plate |
| Product Type | AST Plate |
| Test Type | Antimicrobial Surveillance, Minimum Inhibitory Concentration (MIC) |
| Isolates Per Plate | 1 |
| No. per Pack | 10 |
| Packaging Type | Box |
| Quantity | 10 Plates/Box |
| Reading Options | Manual, Vizion |
| Regulatory Status | For Research Use Only. Not for use in in vitro diagnostic procedures. |
| Species | Enterococcus faecium, Enterococcus faecalis |
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.