missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Semicarbazide Hydrochloride, 99%, (Crystalline), MP Biomedicals
Shop All MP Biomedicals Products
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
Specifications
Specifications
| Assay Percent Range | 99% |
| Quantity | 100 g |
| Formula Weight | 111.5 |
| Physical Form | Crystalline |
| Chemical Name or Material | Semicarbazide Hydrochloride |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction