Learn More
Roetzmeier™ 1.5 in. Self-Closing Tap
Product Code. 10239411
Encompass
Change view
Click to view available options
Material:
Stainless Steel
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material Unit Size
10239411 Stainless Steel Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10239411 Supplier Roetzmeier™ Supplier No. Z150
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
Self-closing tap designed to comfortably and safely dispense precise amounts of liquid.
  • Tap to precisely dispense the amount of liquid you need
  • Self-closing
  • Made of stainless steel 304
  • Gaskets made of PTFE
  • Can be used with all Roetzmeier™ Safety Canisters and Barrels with 1.5 in. opening and separate ventilation
Specifications
Closure Type Tap
For Use With (Equipment) Safety Canisters 05KZ, 10KZ, 20KZ, 20KDL and Safety Transportation Barrel 25TZ
Material Stainless Steel
Quantity 1