Learn More
Selenium AAS Standard, Fluka™
Product Code. 16676232 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16676232 500 mL 500mL
16666232 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 16676232 Supplier Honeywell-Fluka Supplier No. E4067500ML
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Certified Reference Material, 1000 mg/L Se in HNO3 2%
Honeywell Fluka™ analytical standards offers a growing range of high quality inorganic and organic reference materials for the European market produced according to ISO 9001. Our products are suitable for a wide range of industries and applications and come complete with a detailed Certificate of Analysis (CoA) in every delivery, and are eligible for free expert technical support ensuring you have the information you need to quickly start your analysis and pass an audit.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Selenium AAS Standard
Packaging Plastic Bottle
CAS 7783-00-8
pH <2
Melting Point 70°C (Decomposition)
Boiling Point 684.9°C
Quantity 500 mL
Grade AAS