Learn More
Segetalin A Trifluoroacetic Acid Salt, TRC
Specifications
Specifications
| Molecular Weight (g/mol) | 609.7 |
| Quantity | 2.5 mg |
| Shipping Condition | Room Temperature |
| Synonym | Cyclo(L-alanylglycyl-L-valyl-L-prolyl-L-valyl-L-tryptophyl) Trifluoroacetic Acid Salt∣Vaccarin D Trifluoroacetic Acid Salt∣ |