Learn More
3M™ SecureFit™ X5500 Safety Helmet, Vented, CE
Product Code. 17335333
Change view
Click to view available options
Color:
Black
Blue
Green
Hi-vis Green
Orange
Red
Yellow
Unit Size:
Pack of 4
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color unitSize
17335333 Yellow Pack of 4
17345333 Blue Pack of 4
17355333 Green Pack of 4
17365333 Red Pack of 4
17375333 Orange Pack of 4
17385333 Black Pack of 4
17395333 Hi-vis Green Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Product Code. 17335333 Supplier 3M™ Supplier No. 7100175592
  • ONLINE EXCLUSIVE PRICE
    351.00 EUR
    248.80 EUR / Pack of 4
    Save 102.20 EUR
    29% Off
Estimated Shipment: 04-11-2026
Add to basket
Add to basket
3M™ SecureFit™ Safety Helmet with adjustable positioning and easy-to-adjust, 4-point ratchet suspension system that features 3M ™ Pressure Diffusion Technology. 3M™ UVicator™ Sensor indicates over-exposed to UV-radiation.
  • Secure, comfortable head protection against small falling objects and for general industrial use
  • Suspension system incorporates exclusive patented 3M™ Pressure Diffusion Technology
  • 4-point ratchet suspension allows wearer to customise height and fit with multiple levels of vertical adjustment
  • Vents allow airflow through the helmet
  • Optional vents allow airflow through the helmet
  • Inspired by climbing helmets with a brimless design to give you a clear upward view
  • 3M™ Uvicator™ sensor disc tells you when your hard hat has been over exposed to UV-radiation and it's time to replace it
  • 3M™ Scotchlite™ Reflective Material can be added to increase visibility under low light conditions
  • 3M™ Scotchlite™ Reflective Material can also be added to increase visibility under low light conditions
  • Approved for lateral deformation (LD)
  • Approved for lateral deformation
TRUSTED_SUSTAINABILITY
Specifications
Color Yellow
For Use With (Application) Construction, Forestry, Oil, and Gas
Material ABS
Product Type Safety Helmet
Suspension Type 4-point Ratchet
Ventilated Ventilated