Learn More
3M™ SecureFit™ Prescription Insert for 200 and 400 Series, RX-SF400
Product Code. 30784776 Shop All 3M Products
Change view
Click to view available options
Lens Tint:
Clear
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Lens Tint
30784776 Clear
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30784776 Supplier 3M™ Supplier No. 7010528331
  • ONLINE EXCLUSIVE PRICE
    42.00 EUR
    26.94 EUR / Pack of 1
    Save 15.06 EUR
    35% Off
Estimated Shipment: 04-12-2026
Prescription lens inserts are designed to add prescription lenses to 3M™ safety glasses and goggles.
  • Compatible with 3M™ safety glasses and goggles.
  • Holds polycarbonate prescription lenses up to ±6 dioptres.
  • Maintains full eye protection while improving vision.
  • Lightweight and comfortable for extended wear.
TRUSTED_SUSTAINABILITY
Specifications
Lens Coating Uncoated
Lens Tint Clear
Product Type Lens Insert
Frame Color Transparent
Material Plastic
Disposable Reusable
Lens Material Plastic
UV Protection None
Frame Type Frameless