Learn More
3M™ Secure Click™ Half Facepiece Reusable Respirator
Product Code. 16456370
Change view
Click to view available options
Size:
Medium
Large
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
16456370 Medium Each
16466370 Large Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
You may only order up to the available stock. For more details call customer services on +358 9 8027 6280.
Product Code. 16456370 Supplier 3M™ Supplier No. 7100172003
Estimated Shipment: 20-10-2026
Add to basket
Add to basket
You may only order up to the available stock. For more details call customer services on +358 9 8027 6280.
3M™ Secure Click™ Half Facepiece Reusable Respirator is designed with smart and intuitive features, which makes it simple and reliable.
  • Greater breathability and comfort provided by the world's first quad flow cartridge system.
  • Push button seal check helps bolster confidence that the respirator is properly sealed to the face.
  • Speaking diaphragm is designed to help provide easier communication while you work.
  • Confidence that filters and cartridges are installed properly provided by all new Secure Click™ connection.
  • Low profile half mask design offers a wide field of view and compatibility with certain welding and grinding shields.
  • Over-moulded design for easy assembly/disassembly.
  • Advanced silicone material for increased comfort and greater durability.
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance AS/NZS 1716:2012
Connection Type Secure Click™
Color Dark Gray
Material Silicone, Nylon
Product Type Half Facepiece Respirator
Size Medium
Includes Without Speaking Diaphragm
Model HF-802
Quantity 10/Cs.