Learn More
Sebacoyl Dichloride, Sigma-Aldrich™
Product Code. 15820681
Change view
Click to view available options
Quantity:
25 mL
100 mL
Unit Size:
100mL
25mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15820681 100 mL 100mL
15810681 25 mL 25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15820681 Supplier Sigma Aldrich Supplier No. 8188990100
 Restricted Item
Estimated Shipment: 18-11-2026
For synthesis
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 111-19-3
Specifications
Melting Point -5°C
Density 1.12 g/cm3 at 20°C
Boiling Point 161°C/19 hPa
Molecular Formula ClOC(CH2)8COCl
MDL Number MFCD00000770
Quantity 100 mL
Synonym Sebacoyl dichloride, Sebacoyl chloride, Sebacoyl dichloride
Molecular Weight (g/mol) 239.14
Grade Research