Learn More
Eppendorf™ Rotor Lid Sealing.
Product Code. 15294536 Shop All Eppendorf Products
Change view
Click to view available options
For Use With (Equipment):
Rotor lid FA452411 and FA451617
Unit Size:
Pack of 5
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
15294536 Rotor lid FA452411 and FA451617 Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15294536 Supplier Eppendorf™ Supplier No. 5409717006
Estimated Shipment: 12-10-2026
Provides a soft yet secure seal around the edges of a rotor lid.
  • Red in color
  • For FA-45-24-11 and FA-45-16-17 rotor lids
  • 5/pk.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Rotor lid FA452411 and FA451617
Color Red
Product Type Sealing