Learn More
BRAND™ Seal-R-film™ Sealing Film
Product Code. 16435314 Shop All Brand Products
Change view
Click to view available options
Length (Metric):
15 m
38 m
75 m
Width (Metric):
50 mm
100 mm
500 mm
Unit Size:
15m
38m
75m
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Width (Metric) Unit Size
16435314 75 m 50 mm 75m
16425314 38 m 100 mm 38m
16445314 75 m 100 mm 75m
16455314 15 m 500 mm 15m
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 16435314 Supplier BRAND™ Supplier No. 701622
  • ONLINE EXCLUSIVE PRICE
    40.00 EUR
    38.80 EUR / 75m
    Save 1.20 EUR
    3% Off
Estimated Shipment: 12-11-2026
No more spilling, evaporation, and contamination! Seal-R-film™ sealing film seals beakers, flasks, and other vessels. It always fits, even with irregular shapes.
  • The sealing film can be stretched to three times its original length.
  • Seal-R-film™ always fits, even with irregular shapes.
  • For traceability, each roll has a lot identification.
  • With the BRAND cutter for sealing film you can cut rolls with widths of 50 and 100 mm efficiently and economically and store them cleanly.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Film
Width (Metric) 50 mm
Length (Metric) 75 m
Material Mineral Wax, Polyolefin
For Use With (Equipment) Beakers, flasks, and other vessels