Learn More
Hoefer™ SE640 Wide-Mini Vertical Electrophoresis Unit
Product Code. 11985408
Change view
Click to view available options
Type:
Mini Vertical Electrohoresis Unit, Complete
Mini Vertical Electrophoresis Unit, Basic
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Unit Size
11985408 Mini Vertical Electrophoresis Unit, Basic Each
11915358 Mini Vertical Electrohoresis Unit, Complete Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11985408 Supplier Hoefer™ Supplier No. SE640
  • ONLINE EXCLUSIVE PRICE
    1320.00 EUR
    1244.63 EUR / Pack of 1
    Save 75.37 EUR
    5% Off
Estimated Shipment: 04-02-2027
Includes
  • Basic package: Lower buffer chamber (tank), safety lid with leads, upper buffer chamber with cathode and anode electrode fin, 3 sets of glass plates (18 x 8cm), four 8cm clamp assemblies, 6 cams, dual gel casting stand with leveling base and level, buffer dam, Spacer-Mate alignment template, Wonder Wedge plate separation tool, and Gel Seal.
  • Complete package: Lower buffer chamber (tank), safety lid with leads, upper buffer chamber with cathode and anode electrode fin, 3 sets of glass plates (18 x 8cm), four 8cm clamp assemblies, 6 cams, dual gel casting stand with leveling base and level, buffer dam, Spacer-Mate alignment template, Wonder Wedge plate separation tool, Gel Seal, two 15-well 1.5mm combs and two sets of 1.5mm thick (2cm wide) spacers.
Wide-mini system, ideal for rapid screening of multiple samples
The Hoefer Wide-Mini Vertical Electrophoresis unit offers extra wide format ideal for quick, high throughput runs. With accessory divider plate you can complete a run of up to 112 samples in as little as 40 minutes.
  • Maximum glass plate size 18 x 8 cm, permits rapid screening of up to 112 samples at one time
  • Design permits gels to be surrounded uniformly by buffer so heat dissipation is efficient; bands are sharp and lanes are straight
  • Accommodates denaturing or native polyacrylamide gels
  • Run up to four second dimension gels suing IEF strips or tube gels
  • Ideal for optimizing 2DE protocols
  • Storage temperature: +15°C to 30°C
Specifications
Certifications/Compliance EN61010-1, CSA C22.2 1010.1, CE
Description Wide-Mini Vertical Electrophoresis Unit, Basic
Type Mini Vertical Electrophoresis Unit, Basic
Humidity Range Up to 80%
Includes Lower buffer chamber (tank), safety lid with leads, upper buffer chamber with cathode and anode electrode fin, 3 sets of glass plates (18 x 8cm), four 8cm clamp assemblies, 6 cams, dual gel casting stand with leveling base and level, buffer dam, spacer-mate alignment template, wonder wedge plate separation tool and gel seal, bubble level
Temperature (English) Maximum 113°F
Temperature (Metric) Maximum 45°C
Temperature (Metric) Storage 15°C to 30°C
Width (English) 12.59 in.
Width (English) Gel 7.08 in.
Width (Metric) 32 cm
Width (Metric) Gel 18 cm
Height (English) 11.41 in.
Height (English) Gel 3.14 in.
Height (Metric) 29 cm
Depth (English) 5.51 in.
Depth (Metric) 14 cm
Voltage 1000 V
Wattage 50 w
Amperage 500 mA