Learn More
DWK Life Sciences Screw-Top Bottle, Round, Amber, Soda-Lime Glass, High form DIN thread
Shop All DWK Life Sciences ProductsDescription
Round amber soda lime (Type 3) glass bottles, wide mouth and high form thread.
- Amber glass provides UV protection
Specifications
Specifications
| Capacity (Metric) | 2500 mL |
| For Use With (Application) | Storage of light-sensitive materials; Transport and shipping of light-sensitive materials |
| Material | Soda Lime Glass |
| Quantity | 1 Ea. |
| Thread Size (Metric) | GL 45 |
| Diameter (Metric) | 139 mm |
| Autoclavable | Not Autoclavable |