Learn More
Fisherbrand™ Screw Neck Vial, 13-425 Thread, Glass, 4mL
Product Code. 11556044 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Color:
Amber
Clear
Marking Spot:
No
Yes
Unit Size:
Pack of 100
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Marking Spot Unit Size
11556044 Amber No Pack of 100
10571013 Clear No Pack of 100
11576044 Clear Yes Pack of 100
11586044 Amber Yes Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 11556044
Big Savings for Better Separation
Use promo code 27992
View all promotions
In Stock
Add to basket
Edge
Add to basket
Ideal for storage purposes when used with closed top screw seals.
  • Any combination of 4mL Screw Neck Vial ND13 with one of our 13mm PP Screw Seals can be obtained as a 2in1 KIT
  • For storage purposes also available with closed top screw seals
TRUSTED_SUSTAINABILITY
Specifications
Color Amber
Diameter (Metric) Outer 14.7 mm
Material Glass
Volume (Metric) 5 mL
For Use With (Application) HPLC
Neck Style Screw
Packaging Type CleanPack, Polypropylene Box
Volume (Metric) Residual <400 μL
Volume (Metric) Working 4.1 mL
Base Style Flat Bottom
Quantity 100
Closure Size ND13
Marking Spot No
Height (Metric) 45 mm
Product Type 1st Hydrolytic Class