Learn More
Ohaus™ Scout™ SKX Portable Classroom Scale
Product Code. 15569621 Shop All Ohaus Products
Change view
Click to view available options
120 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
15569621 120 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15569621 Supplier Ohaus™ Supplier No. 30253070
Only null left
Add to basket
Add to basket
Designed for education applications and featuring a large backlit LCD and education software.
  • Superior overload protection and practical stackable design for convenient storage.
  • Improved functionality with bright backlit LCD and 4-button operation.
  • Multiple connectivity options available for data logging and communication.
  • Geared for classroom use with education-specific software applications.
TRUSTED_SUSTAINABILITY
Specifications
Connection Type RS232 (Available as an Accessory)USB Host (Available as an Accessory)USB Device (Available as an Accessory)Ethernet (Available as an Accessory)Bluetooth (Available as an Accessory)
Description Electronic Balance, SKX123 EU
Capacity 120 g
Readability 0.001000 g
Includes AC Adapter
Linearity 0.003000 g
Material ABS Housing, Stainless Steel Pan
Model SKX123
Repeatability 0.002000 g
Stabilization Time 1.500 s
Dimensions (L x W x H) 222.00 x 202.00 x 103.000 mm
Length (English) 8.70 in.
Length (Metric) 222.00 mm
Width (English) 8.00 in.
Width (Metric) 202.00 mm
Width (Metric) Pan 93 mm
Height (English) 4.10 in.
Height (Metric) 103.000 mm
Depth (English) 8.70 in.
Depth (Metric) 222.00 mm