Learn More
Kimberly-Clark Professional™ Scott™ Control™ 8042 Folded 2 Ply Toilet Tissue, 36 Packs
Click to view available options
Color:
White
Unit Size:
Pack of 36
Description
- Each individual pack of 2 ply folded toilet tissue contains 250 2 ply toilet paper sheets
- Quality folded toilet paper for where hygiene and contamination control is a priority
- Scott™ Control™ interleaved toilet paper offers single sheet dispensing meaning you only touch the sheets you use helping maximise hygiene and efficiency
- For environmental assurance, commercial toilet paper is made from 100% recycled fibres, is FSC certified, carries the European Ecolabel and has reliable flushability
- Compatible with a variety of Kimberly-Clark toilet paper dispensers
Specifications
Specifications
| Product Type | Folded Toilet Tissue |
| Length (English) | 13.1 in. |
| Length (Metric) | 33.3 cm |
| Width (English) | 11.4 in. |
| Width (Metric) | 29 cm |
| Material | Paper |
| Color | White |
| Description | Folded |
| Packaging Quantity | 250 Sheets/Pk., 36 Pk. |
| Ply | 2-ply |
| Show More |