missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Scopine Hydrochloride, Mikromol™
Description
Scopine Hydrochloride
Specifications
Specifications
| Chemical Name or Material | Scopine Hydrochloride |
| CAS | 85700-55-6 |
| Analyte or Component Names | Scopine Hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C8 H13 N O2 . Cl H |
| Certifications/Compliance | ISO 9001 |
| Synonym | 3-Oxa-9-azatricyclo[3.3.1.02,4]nonan-7-ol, 9-methyl-, hydrochloride (1:1), (1alpha,2beta,4beta,5alpha,7beta)-, 3-Oxa-9-azatricyclo[3.3.1.02,4]nonan-7-ol, 9-methyl-, hydrochloride, (1alpha,2beta,4beta,5alpha,7beta)- (9CI), Scopine hydrochloride |
| InChI Formula | InChI=1S/C8H13NO2.ClH/c1-9-5-2-4(10)3-6(9)8-7(5)11-8;/h4-8,10H,2-3H2,1H3;1H/t4-,5-,6+,7-,8+; |
| SMILES | Cl.CN1[C@@H]2C[C@@H](O)C[C@H]1[C@@H]3O[C@H]23 |
| Molecular Weight (g/mol) | 191.66 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?