Description
- Ag / AgCl reference
- Zero point: pH = 7.0 ± 0.3
- Electrolyte: KCl 3 mol/l
- Pt 1000 temperature sensor
- 1 m long fixed cable
Specifications
Specifications
| Description | Spear membrane |
| Connector Types | BNC |
| Junction | 3 x Platinum |
| Temperature (Metric) Operating | -5°C to +100°C |
| Type | N 5800 BNC |
| Length (Metric) | 96 mm |
| Diameter (Metric) Body | 12 mm |
| Diameter (Metric) Cap | 5 mm |