missing translation for 'onlineSavingsMsg'
Learn More

SCH772984, MedChemExpress

Product Code. 16454027
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16454027 50 mg 50mg
16478108 10 mg 10mg
16427876 100 mg 100mg
16411326 200 mg 200mg
16470568 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16454027 Supplier Medchem Express Supplier No. HY50846
 Restricted Item
Estimated Shipment: 25-09-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress SCH772984 is a highly selective and ATP-competitive ERK inhibitor, with IC50s of 4 and 1 nM for ERK1 and ERK2, respectively. SCH772984 has antitumor activity in MAPK inhibitor-naïve and MAPK inhibitor-resistant cells containing BRAF or RAS mutations.

SCH772984 is a highly selective and ATP-competitive ERK inhibitor, with IC50s of 4 and 1 nM for ERK1 and ERK2, respectively. SCH772984 has antitumor activity in MAPK inhibitor-naïve and MAPK inhibitor-resistant cells containing BRAF or RAS mutations.

Specifications

Chemical Name or Material SCH772984
CAS 942183-80-4
Color Yellow
Purity Grade Notes Research
Molecular Formula C33H33N9O2
Quantity 50 mg
Solubility Information DMSO : 48 mg/mL (81.68 mM; Need ultrasonic)
SMILES O=C([C@H](CC1)CN1CC(N(CC2)CCN2C(C=C3)=CC=C3C4=NC=CC=N4)=O)NC5=CC6=C(C=C5)NN=C6C7=CC=NC=C7
Molecular Weight (g/mol) 587.67
Formula Weight 587.67
Percent Purity 98.69%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.