missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ SCH 51344

Description
SCH 51344 is a potent MTH1 inhibitor (Kd = 49 nM). Inhibits Ras-induced malignant transformation and increases α-actin promoter-driven CAT activity in Ras-transformed cells. Has no effect on Ras-induced ERK and JNK activation. Inhibits Ras-induced membrane ruffling in REF-52 fibroblasts and blocks anchorage-independent growth of Ras-transformed tumor cell lines. Also induces DNA damage in SW480 colon cancer cells.
Specifications
Specifications
| Purity | >99% |
| Quantity | 50 mg |
| Product Type | SCH 51344 |
| Molecular Weight (g/mol) | 316.35 |
| Inhibitors | SCH 51344 |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?