Learn More
Merck Scepter™ Test Bead Vial
Product Code. 17191731
Change view
Click to view available options
Particle Size:
11 μm
Unit Size:
5mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Particle Size Unit Size
17191731 11 μm 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17191731 Supplier Merck Supplier No. PHCC3BEADS
  • ONLINE EXCLUSIVE PRICE
    181.00 EUR
    162.00 EUR / 5mL
    Save 19.00 EUR
    10% Off
Estimated Shipment: 08-02-2027
Scepter™ test beads can be used to assess the Scepter™ cell counter
  • Scepter™ Test Beads can be used to assess the Scepter™ Cell Counter
  • Compatible with Scepter™ 2.0 and 3.0
TRUSTED_SUSTAINABILITY
Specifications
Product Type Test Bead Vial
Particle Size 11 μm
Product Line Scepter
For Use With (Application) Cell Counting