Learn More
Merck Scepter Cell Counter Sensors
Description
- Coupled with precision liquid handling channels and electronics, the Scepter sensor accurately and reliably provides cell population statistics.
- The Scepter 2.0 handheld automated cell counter provides a fast and convenient method for counting cells and particles and is compatible with both the 40 and 60uM sensor. The system employs the Coulter principle in a miniaturized, handheld, format enabling rapid cell counting; what used to take 10 minutes now takes less than 30 seconds. The user prepares a dilution of the cell culture of interest and uses the Scepter cell counter to aspirate a sample of this dilution into the Scepter sensor.
- The Scepter cell counter’s screen displays:
- Cell concentration
- Average cell size
- Average cell volume
- Histogram of size or volume distribution
- This system is intended for research use only and has been tested with cell types representative of those in use today
- 60 μm sensors designed to count particles between 6 μm and 36 μm
- Operating Range: 10,000 particles/mL–500,000 particles/mL
- Partical Size: 6 μm–36 μm
- Cell Size range: 8.0–25.0 μm
Specifications
Specifications
| Includes | 50 μL sample volume |
| Quantity | 250/Pk |