missing translation for 'onlineSavingsMsg'
Learn More

Scandium(III) bis(trifluoromethylsulfonyl)imide

Product Code. p-7080239
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15487345 250 mg 250mg
15462807 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15487345 Supplier Thermo Scientific Alfa Aesar Supplier No. 046948.MD
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 176726-07-1 | C6F18N3O12S6Sc | 885.362 g/mol

Scandium(III) bis(trifluoromethylsulfonyl)imide is used as Lewis acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 176726-07-1
Molecular Formula C6F18N3O12S6Sc
Molecular Weight (g/mol) 885.362
MDL Number MFCD03427000
InChI Key FUXLYEZEIZAKTL-UHFFFAOYSA-N
Synonym scandium iii trifluoromethanesulfonimide
PubChem CID 131875098
IUPAC Name bis(trifluoromethylsulfonyl)azanide;scandium(3+)
SMILES C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F.C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F.C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F.[Sc+3]

Specifications

Melting Point 199°C to 204°C
Color White to Yellow
Linear Formula [(CF3SO2)2N]3Sc
Quantity 250 mg
UN Number UN1759
Sensitivity Moisture sensitive
Solubility Information Insoluble in water.
Formula Weight 885.37
Physical Form Solid
Chemical Name or Material Scandium (III) bis (trifluoromethylsulfonyl)imide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.