missing translation for 'onlineSavingsMsg'
Learn More

SB-3CT, MedChemExpress

Product Code. 16477837
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16477837 5 mg 5mg
16455118 10 mg 10mg
16421357 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16477837 Supplier Medchem Express Supplier No. HY12354
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress SB-3CT is a potent and competitive matrix metalloproteinase MMP-2 and MMP-9 inhibitor with Ki values of 13.9 and 600 nM, respectively. SB-3CT has high selectivity for gelatinases. SB-3CT shows blood-brain barrier permeability and has neuroprotective effects and anticancer activity.

SB-3CT is a potent and competitive matrix metalloproteinase MMP-2 and MMP-9 inhibitor with Ki values of 13.9 and 600 nM, respectively. SB-3CT has high selectivity for gelatinases. SB-3CT shows blood-brain barrier permeability and has neuroprotective effects and anticancer activity.

Specifications

Chemical Name or Material SB-3CT
CAS 292605-14-2
Color Pink
Purity Grade Notes Research
Molecular Formula C15H14O3S2
Quantity 5 mg
Solubility Information DMSO : ≥ 50 mg/mL (163.19 mM)
SMILES O=S(CC1SC1)(C2=CC=C(OC3=CC=CC=C3)C=C2)=O
Molecular Weight (g/mol) 306.4
Formula Weight 306.4
Percent Purity 98.05%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.