missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ SB 24011
missing translation for 'orderingAttributeHoverText'
:
5 mg
Description
SB 24011 is an inhibitor of STING-TRIM29 interaction (IC50 = 3.85 μM in vitro). SB 24011 binds to STING and inhibits TRIM29-E3 ligase interaction to increase cellular STING levels. In syngeneic mouse models, SB 24011 dose-dependently reduces CT26 and B16F10 tumor volumes when co-admistered with cGAMP .
Specifications
Specifications
| Chemical Name or Material | 4-[(2R)-3,4-Dihydro-4-[(4-hydroxyphenyl)methyl]-5-[(4-methoxyphenyl)methyl]-2-(1-methylethyl)-3-oxo-1(2H)-pyrazinyl]-3-nitro-N-[[(2S)-tetrahydro-2-furanyl]methyl]benzamide |
| CAS | 1497415-41-4 |
| Quantity | 5 mg |
| Target | Protein Dimerizers |
| Molecular Formula | C34H38N4O7 |
| Physical Form | Solid |
| Color | Orange |
| Product Type | Bioactive Small Molecule |
| Purity | 0.98 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction