missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ SB 216641 hydrochloride

Description
A selective h5-HT1B antagonist with approximately 25-fold selectivity over h5-HT1D and little or no affinity for a range of other receptor types.
Specifications
Specifications
| Quantity | 10 mg |
| Formula Weight | 523.03 |
| Purity | >98% |
| Molecular Weight (g/mol) | 523.03 |
| Chemical Name or Material | SB 216641 hydrochloride |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?