Learn More
Sartorius MD8 Airport Portable Air Sampler
Product Code. 10638771 Shop All Sartorius Products
Change view
Click to view available options
Airflow Rate:
10, 20, 30, 50, 125 L/min.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Airflow Rate Unit Size
10638771 10, 20, 30, 50, 125 L/min. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10638771 Supplier Sartorius Supplier No. 16757
  • ONLINE EXCLUSIVE PRICE
    6325.00 EUR
    5560.35 EUR / Pack of 1
    Save 764.65 EUR
    12% Off
View all promotions
In Stock
MD8 Airport Portable Air Sampler for impaction measurements and background monitoring with gelatine membranes.
AirPort MD8 is the new air sampler for the pharmaceutical industry, the biotechnology, the food and beverage industry, for hospitals environmental care and for works safety. For guaranteeing reliable and exact measurement results AirPort MD8 uses the gelatine membrane filter method or the Impaction method with BACTair™.
  • Battery-powered and portable for universal use
  • Battery power level clearly indicated so constant performance during sampling is guaranteed
  • Ergonomic design and easy to clean
  • Flexible adjustment possibilities of the volume flow and the sample volume
  • Air Flow:
    • 10 L/min., 20 L/min., 30 L/min., 50 L/min.
    • 125 L/min. (only when BACTairTM culture media plates are used)
  • User friendly prompting with the option of five languages: English, French, German, Italian and Spanish
  • Parameters last used are stored even after automatic shut-off
  • The device can be calibrated on-site
  • CSA Compliant
TRUSTED_SUSTAINABILITY
Specifications
Airflow Rate 10, 20, 30, 50, 125 L/min.
Weight (Metric) 2.5 kg
Battery Rechargeable NiMH
Includes Air Sampler, battery charger, operating instructions, calibration certificate.
Power Consumption 5 to 14 Watt depending of filter pressure drop
Certifications/Compliance CSA Compliant
Product Type Portable Air Sampler
For Use With (Application) For hospitals environmental care and for works safety
Voltage 115/230 V
Dimensions (L x W x H) 300 x 135 x 165 mm