Description
- For hands-free parallel filtration of up to 6 samples
- Permanently connected to vacuum source
- Easily install vacuum filtration units in the MultiStation for quick and easy filtration of samples without the need for installation of extra connectors and time-consuming stabilization
- Works with Sartolab™ RF 50 vacuum filtration units; the funnel adapter of these units is designed to fit perfectly in the bracket of the MultiStation
- With one click, connect the filtration unit to the device, assuring perfect filter stability
- With a second click, engage the vacuum automatically and begin filtering
- Easily manipulate samples with the rotating, multi-directional head
- Easily keep track of samples during filtration with the numbered brackets
- Time-saving (no installation time for each filter unit before use)
Specifications
Specifications
| Autoclavable | Not Autoclavable |
| Color | White |
| Disposable | Reusable |
| Filter Type | Vacuum Filter |
| For Use With (Equipment) | Sartolab RF/BT Units |
| Sterility | Non-sterile |
| For Use With (Application) | Parallel filtration of up to 6 samples |
| Type | Filter Holder |
Product Title
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Ser du en mulighed for forbedring?