Learn More
Sartorius Sartoclear Dynamics™: Lab V
Shop All Sartorius ProductsDescription
Sartoclear Dynamics™ Lab V kits are designed for simultaneous clarification and sterile filtration of 50 mL to 1 L of mammalian cell culture broth to prepare samples for subsequent downstream processing steps, such as protein purification and concentration. The convenient kits combine pouches of filter aid for clarification and vacuum filtration units for sterile filtration.
The Sartoclear Dynamics™ Lab V Kits combine the use of a filter aid for clarification and a vacuum filtration unit with a 0.22 μm polyethersulfone membrane for sterile filtration of up to 1 L mammalian cell cultures. The ready-to-use filter aid is supplied in dust-free and gamma-irradiated pouches.
The Sartolab™ RF bottletop filters with receiver flasks, sterilized by gamma irradiation are
available in volume capacities ranging from 150 mL to 1,000 mL.
- Clarify high-density mammalian cell culture in minutes
- Out-of-the-box solution
- Fast and effortless
- No need for centrifugation
Ten kits are available to meet all your needs based on your
filtration volume and the cell density of your mammalian
cell cultures.
Specifications
Specifications
| Filter Type | Diatomaceous Earth/Heavy metal-free polystyrene/Heavy metal-free polyethylene |
| Product Type | Sartoclear Dynamics&trade: Lab V kits have been designed for cell harvesting of mammalian cell cultures, such as CHO, HEK, hybridomas and many others, with high cell densities. |
| Bed Volume | 1,000mL |
| Quantity | 12 x 1L Filter Units + 40g Filter Aid |
| Flow Rate | 0.22 μm polyethersulfone/high flow rate |
| Includes | Each Sartoclear Dynamics&trade: Lab V kit is comprised of pouches of filter aid and vacuum filtration units (Sartolab&trade: RF) that match your needs. Find the right kit in just two easy steps: 1. Determine the volume range of your sample to be filtered. 2. Then reference this range to the cell density of your cell culture |
| Content And Storage | ambient |
| For Use With (Application) | mammalian cell harvesting/culture |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.