Learn More
Sarcosine, Sigma-Aldrich™
Specifications
Specifications
| CAS | 107-97-1 |
| MDL Number | MFCD00004279 |
| Quantity | 250 g |
| Synonym | Sarcosine, Methylaminoacetic acid, N-Methylglycine, Sarcosine |
| Molecular Weight (g/mol) | 89.09 |
| Grade | Research |