missing translation for 'onlineSavingsMsg'
Learn More

Sarcosine Ethyl Ester Hydrochloride, TRC

Product Code. 30411115
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30411115 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30411115 Supplier Toronto Research Chemicals Supplier No. TRCS14051010G
 Restricted Item
Estimated Shipment: 11-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Sarcosine Ethyl Ester Hydrochloride
CAS 52605-49-9
Molecular Formula C5H12ClNO2
Quantity 10 g
Synonym (Methylamino)acetic acid ethyl ester hydrochloride, 2-(Methylamino)acetic acid ethyl ester hydrochloride, 2-Ethoxy-2-oxo-N-methylethylammonium chloride, Ethyl N-methylaminoacetate hydrochloride, Ethyl N-methylglycinate hydrochloride, Ethyl sarcosinate hydrochloride, N-Methylglycine ethyl ester hydrochloride, N-[(Ethoxycarbonyl)methyl]-N-methylamine hydrochloride, Sacrosine ethyl ester hydrochloride, Sarcosine ethyl ester hydrochloride
InChI Formula InChI=1S/C5H11NO2.ClH/c1-3-8-5(7)4-6-2;/h6H,3-4H2,1-2H3;1H
SMILES O=C(OCC)CNC.[H]Cl
IUPAC Name ethyl methylglycinate hydrochloride
Molecular Weight (g/mol) 153.61
Formula Weight 153.06
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.