missing translation for 'onlineSavingsMsg'
Learn More

SAR405838, MedChemExpress

Product Code. p-200100082
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16470138 5 mg 5mg
16457678 10 mg 10mg
16437576 100 mg 100mg
16423687 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16470138 Supplier Medchem Express Supplier No. HY18986

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress SAR405838 (MI-77301), an analog of MI-773, is a highly potent and selective MDM2-p53 interaction inhibitor. SAR405838 binds to MDM2 with a Ki of 0.88 nM. SAR405838 induces apoptosis and has potent antitumor activity.

SAR405838 (MI-77301), an analog of MI-773, is a highly potent and selective MDM2-p53 interaction inhibitor. SAR405838 binds to MDM2 with a Ki of 0.88 nM. SAR405838 induces apoptosis and has potent antitumor activity.

Specifications

Chemical Name or Material SAR405838
CAS 1303607-60-4
Color Off-White
Purity Grade Notes Research
Molecular Formula C29H34Cl2FN3O3
Quantity 5 mg
Synonym MI-77301
Solubility Information DMSO : 100 mg/mL (177.78 mM; Need ultrasonic)
SMILES O=C([C@H](N[C@H]1CC(C)(C)C)[C@H](C2=CC=CC(Cl)=C2F)[C@@]31C(NC4=C3C=CC(Cl)=C4)=O)N[C@@H]5CC[C@@H](O)CC5
Molecular Weight (g/mol) 562.5
Formula Weight 562.5
Percent Purity 95.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.