Learn More
(-)-Santonin, TRC
Product Code. 30416501
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30416501 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30416501 Supplier Toronto Research Chemicals Supplier No. TRCS115300250MG
 Restricted Item
Estimated Shipment: 10-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material alpha-Santonin
CAS 481-06-1
Molecular Formula C15 H18 O3
Quantity 250 mg
Synonym (3S,3aS,5aS,9bS)-3a,5,5a,9b-tetrahydro-3,5a,9-trimethylnaphtho[1,2-b]furan-2,8(3H,4H)-dione, (11S)-6α-hydroxy-3-oxo-eudesma-1,4-dien-12-oic acid γ-lactone, [3S-(3α,3aα,5aβ,9bβ)]-3a,5,5a,9b-tetrahydro-3,5a,9-trimethylnaphtho[1,2-b]furan-2,8(3H,4H)-dione, (-)-Santonin, (-)-α-Santonin, 1,2,3,4,4a,7-Hexahydro-1-hydroxy-α,4a,8-trimethyl-7-oxo-2-naphthaleneacetic acid γ-lactone, NSC 4900, Santonin, Semenen, alpha-Santonin, α-Santonin
InChI Formula InChI=1S/C15H18O3/c1-8-10-4-6-15(3)7-5-11(16)9(2)12(15)13(10)18-14(8)17/h5,7-8,10,13H,4,6H2,1-3H3/t8-,10-,13-,15-/m0/s1
SMILES C[C@H]1[C@@H]2CC[C@@]3(C)C=CC(=O)C(=C3[C@H]2OC1=O)C
IUPAC Name (3S,3aS,5aS,9bS)-3,5a,9-trimethyl-3a,4,5,9b-tetrahydro-3H-benzo[g][1]benzofuran-2,8-dione
Molecular Weight (g/mol) 246.3
Formula Weight 246.1256