Learn More
Dräger™ Sampling Hoses and Tools for Dräger™ accuro™ Bellows Pumps
Product Code. 12857820
Change view
Click to view available options
Length (Metric):
3 m
10 m
15 m
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) unitSize
12857820 3 m Each
12867820 10 m Each
15568601 15 m Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 12857820 Supplier Dräger™ Supplier No. 6400077
  • ONLINE EXCLUSIVE PRICE
    280.00 EUR
    254.51 EUR / Pack of 1
    Save 25.49 EUR
    9% Off
Estimated Shipment: 08-09-2026
Specifications
For Use With (Equipment) Accuro Bellows Pump
Type Extension Hose
Includes Adapter
Length (English) 10 ft.
Length (Metric) 3 m