Learn More
Buerkle™ Angular Beaker for use with Buerkle™ Telescopic Rod
Description
- Polypropylene
- For industry and water treatment
Specifications
Specifications
| Product Type | Water Sampler |
| Material | Polypropylene |
| Autoclavable | Not Autoclavable |
| Capacity (English) | 33.81 oz. |
| Capacity (Metric) | 1000 mL |
| Style | Angular |
| For Use With (Application) | For Industry and Water Treatment |