Learn More
S Murray and Co™ SAMCO™ Stainless Steel Scissors
Product Code. 12982055
Change view
Click to view available options
Length (Metric):
115 mm
125 mm
150 mm
200 mm
Blade Style:
Curved
Straight
Tip Style:
1 Pointed, 1 Blunt
Fine Point
Very Fine Point
Unit Size:
Each
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Blade Style Tip Style Unit Size
12982055 115 mm Curved Very Fine Point Each
12358099 125 mm Straight 1 Pointed, 1 Blunt Each
12348099 115 mm Straight Very Fine Point Each
12912055 125 mm Straight Fine Point Each
12972055 125 mm Straight Very Fine Point Each
12494070 150 mm Straight Fine Point Each
12932055 200 mm Straight Very Fine Point Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
You may only order up to the available stock. For more details call customer services on +358 9 8027 6280.
Product Code. 12982055 Supplier S Murray and Co™ Supplier No. E830/01
  • ONLINE EXCLUSIVE PRICE
    7.40 EUR
    4.12 EUR / Pack of 1
    Save 3.28 EUR
    44% Off
Estimated Shipment: 22-12-2026
You may only order up to the available stock. For more details call customer services on +358 9 8027 6280.
Use these scissors for a wide range of applications. S Murray™ SAMCO™ Stainless Steel Scissors are available in a variety of blade and tip styles to meet your needs.
  • Fully guaranteed in normal use and subject to stringent quality inspection procedures
Specifications
Blade Style Curved
Length (Metric) 115 mm
Material Stainless Steel
Description Very fine points and closed shanks
Tip Style Very Fine Point
Product Type Stainless Steel Scissors