Learn More
S Murray and Co™ SAMCO™ Household Scissors
Product Code. 12932065
Change view
Click to view available options
Length (Metric):
150 mm
175 mm
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) unitSize
12932065 150 mm Each
12942065 175 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 12932065 Supplier S Murray and Co™ Supplier No. E901/04
Estimated Shipment: 11-09-2026
Add to basket
Add to basket
Use these scissors for cutting anything from bandages to stainless steel mesh. S Murray™ SAMCO™ Household Scissors are heavy duty but lightweight with stainless steel blades and robust construction.
  • General purpose, nickel plated steel household scissors
  • Straight with one sharp point
  • Autoclavable
Specifications
Product Type Household Scissors
Blade Style Straight
Length (Metric) 150 mm
Material Stainless Steel