missing translation for 'onlineSavingsMsg'
Learn More

Samarium, Sigma-Aldrich™

Product Code. 15869940
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15869940 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15869940 Supplier Sigma Aldrich Supplier No. 8144790001
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Powder for synthesis

TRUSTED_SUSTAINABILITY

Specifications

CAS 7440-19-9
Melting Point 1072°C
Quantity 1 g
Form Powder
MDL Number MFCD00011233
Synonym Samarium, Samarium
Molecular Weight (g/mol) 150.36
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.