Learn More
Samarium, Sigma-Aldrich™
Specifications
Specifications
| Chemical Name or Material | Samarium |
| CAS | 7440-19-9 |
| Melting Point | 1072°C |
| Quantity | 10 g |
| Synonym | Samarium, Samarium |
| Molecular Weight (g/mol) | 150.36 |