missing translation for 'onlineSavingsMsg'
Learn More

Samarium(III) Oxide, Sigma-Aldrich™

Product Code. 15847910
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15847910 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15847910 Supplier Sigma Aldrich Supplier No. 1123210010
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Melting Point 2325°C
CAS 12060-58-1
Quantity 10 g
MDL Number MFCD00011237
Synonym Samarium(III) oxide, Samarium(III) oxide
Molecular Weight (g/mol) 348.72
Grade Research
Density 8.347 g/cm3
Chemical Name or Material Samarium(III) Oxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.