missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Salvinorin B
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
Description
Salvinorin B is a potent and selective κ-opioid DREADD (KORD) activator (EC50 = 11.8 nM). Selective for KORD over endogenous κ opioid receptor and a range of other related targets. Exhibits no analgesic or ataxic effects in wild type mice. Induces neuronal hyperpolarization, and modifies locomotor activity and feeding behavior in KORD-expressing mouse models. Brain penetrant. Metabolite of salvinorin A.
Specifications
Specifications
| Chemical Name or Material | (2S,4aR,6aR,7R,9S,10aS,10bR)-2-(3-Furanyl)dodecahydro-9-hydroxy-6a,10b-dimethyl-4,10-dioxo-2H-naphtho[2,1-c]pyran-7-carboxylic acid methyl ester |
| CAS | 92545-30-7 |
| Quantity | 1 mg |
| Target | DREADD Activators |
| Molecular Formula | C21H26O7 |
| Purity | 98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction