Learn More
Salmeterol Dimer Impurity (Mixture of Diastereomers), TRC
Specifications
Specifications
| Chemical Name or Material | Salmeterol Dimer Impurity (Mixture of Diastereomers) |
| CAS | 2470130-36-8 |
| Molecular Formula | C50H72N2O7 |
| Quantity | 2.5 mg |
| Synonym | 1-[4-Hydroxy-3-[[[2-hydroxy-2-[4-hydroxy-3-(hydroxymethyl)phenyl]ethyl][6-(4-phenylbutoxy)hexyl]amino]methyl]phenyl]2-[[6-(4-phenylbutoxy)hexyl]amino]ethanol, Salmeterol EP Impurity G |
| InChI Formula | InChI=1S/C50H72N2O7/c53-40-46-36-44(26-28-48(46)55)50(57)39-52(30-14-2-4-16-32-59-34-18-12-24-42-21-9-6-10-22-42)38-45-35-43(25-27-47(45)54)49(56)37-51-29-13-1-3-15-31-58-33-17-11-23-41-19-7-5-8-20-41/h5-10,19-22,25-28,35-36,49-51,53-57H,1-4,11-18,23-24,29-34,37-40H2 |
| SMILES | OC1=CC=C(C=C1CO)C(O)CN(CC2=CC(=CC=C2O)C(O)CNCCCCCCOCCCCC=3C=CC=CC3)CCCCCCOCCCCC=4C=CC=CC4 |
| IUPAC Name | 4-(1-hydroxy-2-((2-hydroxy-5-(1-hydroxy-2-((6-(4-phenylbutoxy)hexyl)amino)ethyl)benzyl)(6-(4-phenylbutoxy)hexyl)amino)ethyl)-2-(hydroxymethyl)phenol |
| Molecular Weight (g/mol) | 813.12 |
| Formula Weight | 812.53 |