Learn More
Saint Gobain Life Sciences Tygon™ ND 100-65 Medical Tubing
Product Code. 10366933
Change view
Click to view available options
Diameter (English) Inner:
0.375 in.
0.5 in.
1/16 in.
1/4 in.
3/16 in.
3/32 in.
5/16 in.
Diameter (English) Outer:
0.375 in.
0.5 in.
0.563 in.
1/8 in.
11/16 in.
5/16 in.
5/32 in.
7/16 in.
Thickness (English):
1/16 in.
1/32 in.
1/8 in.
3/32 in.
Unit Size:
15m
Each
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (English) Inner Diameter (English) Outer Thickness (English) unitSize
10366933 1/16 in. 1/8 in. 1/32 in. 15m
10328184 3/32 in. 5/32 in. 1/32 in. 15m
12356839 3/16 in. 5/16 in. 1/16 in. Each
10667925 1/4 in. 0.375 in. 1/16 in. 15m
12396839 1/4 in. 7/16 in. 3/32 in. 15m
12316849 5/16 in. 7/16 in. 1/16 in. Each
12306849 1/4 in. 0.5 in. 1/8 in. Each
12346849 0.375 in. 0.5 in. 1/16 in. Each
12356849 0.375 in. 0.563 in. 3/32 in. Each
15692054 0.5 in. 11/16 in. 3/32 in. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Product Code. 10366933 Supplier Saint Gobain Life Sciences Supplier No. ADF00002
Add to basket
Add to basket
Tygon™ ND 100-65 Medical Tubing for blood contact medical devices where non-DEHP plasticized materials are required.
  • Crystal clear for easy visualization of fluid flow
  • For contact with bodily fluids
  • Non-wetting surface permits complete drainage
  • Non-hemolytic and non-pyrogenic
  • Withstands EtO, gas and gamma sterilization
  • Minimally invasive devices
  • Blood and IV solutions
  • Dialysis equipment
  • Wound drainage
  • Inhalation equipment
  • Chemotherapy drug delivery
  • Kidney dialysis pumps
  • Peristaltic pumps
  • Catheters
Compliance
Meets USP Class VI criteria, USP Class VI, ISO 10993, European Pharmacopoeia 3.1.1.2 and European REACH requirements.
TRUSTED_SUSTAINABILITY
Specifications
Diameter (English) Inner 1/16 in.
Diameter (Metric) Inner 1.6 mm
Material Non-DEHP Plastic
Diameter (English) Outer 1/8 in.
Diameter (Metric) Outer 3.2 mm
Overall Length (English) 50 ft.
Overall Length (Metric) 15.2 m
Thickness (English) 1/32 in.
Thickness (Metric) 0.8 mm
Color Clear
Product Type Tubing