Learn More
Saflufenacil-d7, TRC
Product Code. 30424614
Change view
Click to view available options
Quantity:
0.5 mg
1 mg
Unit Size:
0.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30424614 1 mg -
30417560 0.5 mg 0.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30424614 Supplier Toronto Research Chemicals Supplier No. TRCS0818021MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 2469260-39-5
Molecular Formula C17H10D7ClF4N4O5S
Molecular Weight (g/mol) 507.9
Synonym 2-Chloro-5-[3,6-dihydro-3-methyl-2,6-dioxo-4-(trifluoromethyl)-1(2H)-pyrimidinyl]-4-fluoro-N-[[methyl(1-methylethyl)amino]sulfonyl]benzamide-d7, 2-Chloro-4-fluoro-N-[isopropyl(methyl)sulfamoyl]-5-[3-methyl-2,6-dioxo-4-(trifluoromethyl)pyrimidin-1-yl]benzamide-d7, 2-Chloro-5-[3,6-dihydro-3-methyl-2,6-dioxo-4-(trifluoromethyl)-1(2H)-pyrimidinyl]-4-fluoro-N-[(isopropyl)methylsulfamoyl]benzamide-d7, 2-Chloro-5-[3,6-dihydro-3-methyl-2,6-dioxo-4-trifluoromethyl-1(2H)-pyrimidinyl]-4-fluoro-N-[[N-methyl-N-(1-methylethyl)amino]sulfonyl]benzamide-d7, Benmihuangcaoan-d7, Eragon-d7, Kixor-d7, N'-[2-Chloro-4-fluoro-5-[1,2,3,6-tetrahydro-3-methyl-2,6-dioxo-4-(trifluoromethyl)pyrimidin-1-yl]benzoyl]-N-isopropyl-N-methylsulfamide-d7, Sharpen-d7, Treevix-d7
IUPAC Name 2-chloro-4-fluoro-5-(3-methyl-2,6-dioxo-4-(trifluoromethyl)-3,6-dihydropyrimidin-1(2H)-yl)-N-(N-methyl-N-(propan-2-yl-d7)sulfamoyl)benzamide
SMILES O=C(NS(=O)(N(C([2H])(C([2H])([2H])[2H])C([2H])([2H])[2H])C)=O)C1=CC(N(C(N(C)C(C(F)(F)F)=C2)=O)C2=O)=C(F)C=C1Cl
Specifications
Quantity 1 mg
InChI Formula InChI=1S/C17H17ClF4N4O5S/c1-8(2)25(4)32(30,31)23-15(28)9-5-12(11(19)6-10(9)18)26-14(27)7-13(17(20,21)22)24(3)16(26)29/h5-8H,1-4H3,(H,23,28)/i1D3,2D3,8D
Formula Weight 507.1
Chemical Name or Material Saflufenacil-d7